Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SF1 Monoclonal Antibody (2F5D10)
GREENER_CHOICE

Catalog No. PIMA549179
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549179 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549179 Supplier Invitrogen™ Supplier No. MA549179
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human SKOV-3 whole cell, human K562 whole cell, human THP-1 whole cell, human A549 whole cell, rat PC-12 whole cell, mouse NIH/3T3 whole cell. IHC: human bladder cancer tissue, human breast cancer tissue, human liver cancer tissue, human placenta tissue, human squamous metaplasia of the renal pelvis tissue, mouse colon tissue, mouse kidney tissue, rat colon tissue, rat kidney tissue. ICC/IF: A431 cell. Flow: A431 cell, Neuro-2a cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Splicing factor 1 is a nuclear pre-mRNA splicing factor. The protein specifically recognizes the intron branch point sequence at the 3' splice site, together with the large subunit of U2 auxiliary factor (U2AF), and is required for the early stages of spliceosome assembly. It also plays a role in nuclear pre-mRNA retention and transcriptional repression. The protein contains an N-terminal U2AF ligand motif, a central hnRNP K homology motif and quaking 2 region which bind a key branch-site adenosine within the branch point sequence, a zinc knuckles domain, and a C-terminal proline-rich domain. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2F5D10
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene SF1
Gene Accession No. Q15637, Q64213
Gene Alias BBP; D11S636; M ZFM1; Mammalian branch point-binding protein; MBBP; MZFM; SF; SF1; Splicing factor 1; transcription factor ZFM1; WBP4; wu:fc09f06; ZCCHC25; ZFM1; Zfp162; Zinc finger gene in MEN1 locus; zinc finger protein 162; ZNF162
Gene Symbols SF1
Host Species Mouse
Immunogen E. coli-derived human splicing factor 1 recombinant protein (Position: R160-Q266).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117855, 22668, 7536
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.