Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SF3A3 Recombinant Protein

Catalog No. 89012090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-012-090 25 μg
89-012-089 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-012-090 Supplier Abnova™ Supplier No. H00010946P0125
Add to Cart
Add to Cart

Human SF3A3 full-length ORF recombinant protein with GST-tag at N-terminal

This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20.

  • Theoretical MW (kDa): 80.85
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: METILEQQRRYHEEKERLMDVMAKEMLTKKSTLRDQINSDHRTRAMQDRYMEVSGNLRDLYDDKDGLRKEELNAISGPNEFAEFYNRLKQIKEFHRKHPNEICVPMSVEFEELLKARENPSEEAQNLVEFTDEEGYGRYLDLHDCYLKYINLKASEKLDYITYLSIFDQLFDIPKERKNAEYKRYLEMLLEYLQDYTDRVKPLQDQNELFGKIQAEFEKKWENGTFPGWPKETSSALTHAGAHLDLSAFSSWEELASLGLDRLKSALLALGLKCGGTLEERAQRLFSTKGKSLESLDTSLFAKNPKSKGTKRDTERNKDIAFLEAQIYEYVEILGEQRHLTHENVQRKQARTGEEREEEEEEQISESESEDEENEIIYNPKNLPLGWDGKPIPYWLYKLHGLNINYNCEICGNYTYRGPKAFQRHFAEWRHAHGMRCLGIPNTAHFANVTQIEDAVSLWAKLKLQKASERWQPDTEEEYEDSSGNVVNKKTYEDLKRQGLL

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH11523.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 10946
Molecular Weight (g/mol) 80.85
Name SF3A3 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen METILEQQRRYHEEKERLMDVMAKEMLTKKSTLRDQINSDHRTRAMQDRYMEVSGNLRDLYDDKDGLRKEELNAISGPNEFAEFYNRLKQIKEFHRKHPNEICVPMSVEFEELLKARENPSEEAQNLVEFTDEEGYGRYLDLHDCYLKYINLKASEKLDYITYLSIFDQLFDIPKERKNAEYKRYLEMLLEYLQDYTDRVKPLQDQNELFGKIQAEFEKKWENGTFPGWPKETSSALTHAGAHLDLSAFSSWEELASLGLDRLKSALLALGLKCGGTLEERAQRLFSTKGKSLESLDTSLFAKNPKSKGTKRDTERNKDIAFLEAQIYEYVEILGEQRHLTHENVQRKQARTGEEREEEEEEQISESESEDEENEIIYNPKNLPLGWDGKPIPYWLYKLHGLNINYNCEICGNYTYRGPKAFQRHFAEWRHAHGMRCLGIPNTAHFANVTQIEDAVSLWAKLKLQKASERWQPDTEEEYEDSSGNVVNKKTYEDLKRQGLL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias PRP9/PRPF9/SAP61/SF3a60
Common Name SF3A3
Gene Symbol SF3A3
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.