Learn More
SF3b155/SAP155 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Specifications
Specifications
| Antigen | SF3b155/SAP155 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | SF3B1 |
| Gene Accession No. | O75533 |
| Gene Alias | Hsh155, PRP10, PRPF10, SAP155, SF3b155, SAP 155; pre-mRNA processing 10; pre-mRNA splicing factor SF3b, 155 kDa subunit; pre-mRNA-splicing factor SF3b 155 kDa subunit; spliceosome associated protein 155; spliceosome-associated protein 155; splicing factor 3B subunit 1 |
| Gene Symbols | SF3B1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.