Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFMBT1 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C396)
GREENER_CHOICE

Catalog No. PIMA548339
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548339 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548339 Supplier Invitrogen™ Supplier No. MA548339
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody recognizes SFMBT1 (Scm-like with four MBT domains protein 1). It binds to a folded domain, amino acids 1-455. SFMBT1 is a histone-binding protein, which is part of various corepressor complexes. It mediates the recruitment of corepressor complexes to target genes, followed by chromatin compaction and repression of transcription and plays a role during myogenesis.

This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFMBT1 Chimeric
Applications ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RAB-C396
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene SFMBT1
Gene Accession No. Q9UHJ3
Gene Alias 4930442N21Rik; 9330180L21Rik; AA536974; hSFMBT; Renal ubiquitous protein 1; RU1; Scm like with four mbt domains 1; Scm-like with four mbt domains 1; scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene containing four mbt domains 1; Scm-related gene product containing four mbt domains; SFMBT; SFMBT1; Smr; wu:fc12f05
Gene Symbols SFMBT1
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing SFMBT1 protein under non-denaturing conditions.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51460
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.