Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529390 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529390 Supplier Invitrogen™ Supplier No. PA529390
  • $586.00
    $478.65 / Each of 1

    Save $107.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLa. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Dog (100%), Pig (100%), Sheep (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SFRP2
Gene Accession No. Q96HF1
Gene Alias AI851596; FKSG12; FRP 2; FRP2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP1; SARP-1; SDF 5; Sdf5; SDF-5; secreted apoptosis related protein 1; secreted apoptosis-related protein 1; secreted frizzled related protein 2; secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; SFRP2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170; UNQ361/PRO697
Gene Symbols SFRP2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SFRP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.