Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFRP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571770
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571770 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571770 Supplier Invitrogen™ Supplier No. PA571770
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SFRP5 Polyclonal Antibody for IHC (P), Western Blot

Secreted frizzled-related protein 5 (SFRP5) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP5 and SFRP1 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP5 is highly expressed in the retinal pigment epithelium, and moderately expressed in the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SFRP5
Gene Accession No. Q5T4F7, Q9WU66
Gene Alias AI605071; Frizzled-related protein 1b; FRP1B; FRP-1b; SARP3; SARP-3; secreted apoptosis related protein 3; secreted apoptosis-related protein 3; secreted frizzled related protein 5; secreted frizzled-related protein 5; secreted frizzled-related sequence protein 5; SFRP5; sFRP-5
Gene Symbols SFRP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 190-219 amino acids from the Central region of human SFRP5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54612, 6425
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.