Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS7/9G8/SRp20 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1642 Supplier Bioss Supplier No. BS11755R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.

Specifications

Antigen SFRS7/9G8/SRp20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Srsf7
Gene Accession No. Q16629
Gene Alias 35kDa; 9430065L19Rik; 9G8; AAG3; aging-associated protein 3; BOS_11471; NX-96; serine and arginine rich splicing factor 7; Serine/arginine-rich splicing factor 7; serine/arginine-rich splicing factor 7-like protein; Sfrs7; Splicing factor 9G8; Splicing factor, arginine/serine-rich 7; splicing factor, arginine/serine-rich 7, 35kDa; SR splicing factor 7; Srsf7
Gene Symbols Srsf7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SFRS7.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6432
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.