Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SFRS8/SWAP Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140561
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8952 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8952 Supplier Bethyl Laboratories, Inc Supplier No. A300985AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections.

SWAP (suppressor of white apricot protein homolog), also known as SFRS8 (splicing factor, arginine/serine-rich 8), is the 951 amino acid human homolog of a Drosophilia splicing protein. Localized to the nucleus, SWAP contains two SURP repeats through which it is thought to mediate splicing events, possibly regulating the alternative splicing of Fibronectin and CD45RC. SWAP regulates its own expression levels (via control of splicing in its first two introns) and may act in tandem with other arginine/serine-rich splicing factors to control protein expression. The gene encoding SWAP is located on a region of chromosome 12 that is related to asthma susceptibility, possibly indicating a role for SWAP in the development of asthma. Multiple isoforms of SWAP exist due to alternative splicing events.

Specifications

Antigen SFRS8/SWAP
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SFSWAP
Gene Accession No. Q12872
Gene Alias Splicing factor, suppressor of white-apricot homolog, Splicing factor, arginine/serine-rich 8, Suppressor of white apricot protein homolog, SFSWAP, SFRS8, SWAP
Gene Symbols SFSWAP
Host Species Rabbit
Immunogen Between 500 and 550
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6433
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.