Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPB Monoclonal Antibody (SPB02 (1B9))
GREENER_CHOICE

Catalog No. PIMA1204 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1204 Supplier Invitrogen™ Supplier No. MA1204
  • $623.00
    $449.65 / Each of 1

    Save $173.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Detects mature surfactant protein B (8kDa monomer and 16kDa Dimeric forms) and pro surfactant protein B (24kDa) in western blot. Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 92%; Rabbit: 85%; Rat: 100%; Sheep: 79%.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPB
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone SPB02 (1B9)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene SFTPB
Gene Accession No. P07988
Gene Alias 18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B
Gene Symbols SFTPB
Host Species Mouse
Immunogen Recombinant Surfactant Protein B.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6439
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.