Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595506
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595506 Supplier Invitrogen™ Supplier No. PA595506
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell, human COLO-320 whole cell, human HepG2 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventilation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPB
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene SFTPB
Gene Accession No. P07988, P22355, P50405
Gene Alias 18 kDa pulmonary-surfactant protein; 6 kDa protein; AI562151; P SFTB3; pot. surfactant-associated protein precursor; putative; preproprotein; PSP-B; Pulmonary surfactant protein 18; pulmonary surfactant protein B; pulmonary surfactant-associated protein B; pulmonary surfactant-associated protein B precursor; pulmonary surfactant-associated protein B; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein B; Pulmonary surfactant-associated proteolipid SPL(Phe); SF-B; SFPB; SFTB3; SFTP3; Sftp-3; Sftpb; SMDP1; SP 18; SP-B; SPBS; surfactant associated protein B; surfactant protein B; surfactant protein-B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B
Gene Symbols SFTPB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human SFTPB (QCLAERYSVILLDTLLGRMLPQLVCRLVLR).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192155, 20388, 6439
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.