Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SFTPD Monoclonal Antibody (VIF9)
GREENER_CHOICE

Catalog No. 01676580 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-580 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-580 Supplier Invitrogen™ Supplier No. MA140247
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-40247 detects SP-D from rat and human samples. MA1-40247 has been successfully used in ELISA, Western blot and immunohistochemistry (paraffin) applications. The MA1-40247 immunogen is rat SP-D.

Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VIF9
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene SFTPD
Gene Accession No. P35247, P35248
Gene Alias AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Gene Symbols SFTPD
Host Species Mouse
Immunogen Rat SP-D.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25350, 6441
Target Species Human, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.