Learn More
ACROBiosystems SFTS virus pre-Gc protein 50ug
Supplier:  ACROBiosystems PRNS52H350UG
Encompass_Preferred
SFTS virus pre-Gc Protein (HN6) His Tag (PRN-S52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 563 - Arg 1035 (Accession # F1BWU4).
Catalog No. 50-253-4286
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.