Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SFX-Insect™ Cell Culture Media

Catalog No. SH3035003 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188
Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Phenol Red, No HEPES, No Protein
No Protein, No HEPES
No Protein, No Phenol Red, No HEPES
No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES
Form:
Liquid
Powder
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity With Additives Without Additives Form
SH3035003 10 L Glutamine, Poloxamer 188 No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES Powder
SH30278LS 6 x 1000 mL Glutamine, Sodium Bicarbonate, Poloxamer 188 No Protein, No Phenol Red, No HEPES Liquid
SH3035002 5 L Glutamine, Poloxamer 188 No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES Powder
SH3027802 1000 mL Glutamine, Poloxamer 188 No Protein, No Phenol Red, No HEPES Liquid
SH3027806 10 L Glutamine, Sodium Bicarbonate, Poloxamer 188 No Protein, No HEPES Liquid
SH3027801 500 mL Glutamine, Poloxamer 188 No Phenol Red, No HEPES, No Protein Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH3035003 Supplier Cytiva Supplier No. SH30350.03
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to support the growth of multiple insect cell lines and production of a variety of recombinant proteins. HyClone™ SFX Insect™ Cell Culture Media is a protein-free media that support cell culture growth.

  • Protein free media
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Insect Cell Medium
Sterility Non-sterile
Sterilization Method Non-sterile
With Additives Glutamine, Poloxamer 188
Without Additives No Sodium Bicarbonate, No Protein, No Phenol Red, No HEPES
Quantity 10 L
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light and moisture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.