Learn More
SGC 0946, Tocris Bioscience™
Description
To request the negative control for SGC0946, please fill out the SGC0649 request form on the SGC website.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 618.57 |
| Percent Purity | >99% |
| Chemical Name or Material | SGC 0946 |
Safety and Handling
| Recommended Storage | Store at -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.