Learn More
TARGETMOL CHEMICALS INC SGC-CBP30 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6668
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SGC-CBP30 is an effective CREBBP/EP300 inhibitor (IC50 21/38 nM). purity: 99%
Catalog No. 50-257-6476
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.