Learn More
Active Motif SGC707
SDP
Supplier:  Active Motif 14130
Encompass_Preferred
Active Motif's SGC707 is a potent and selective allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3), IC50 = 31 nM and selective against 31 other methyltransferases. SGC707 is an extremely useful probe for investigating the role of PRMT3 in normal and pathophysiology. it is cell permeable and active in vivo.
Catalog No. NC1841456
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.