Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc SGE 100% Graphite Ferrule

Catalog No. 502395830 Shop All Phenomenex Products
Encompass_Preferred

SGE 100% Graphite Ferrule 1/8in Nut, 0.5mm ID

Catalog No. 50-239-5830 Supplier Phenomenex Inc Supplier No. 072624
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SGE 100% Graphite Ferrule 1/8in Nut, 0.5mm ID

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.