Learn More
MilliporeSigma™ Supelco™ SGE Autosampler Syringe, Fixed Needle (FN)
Catalog No. 111008891 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
Quantity:
6 Pk.
Catalog No. Volume (Metric) Quantity
11-100-8891 10 μL 6 Pk.
1 options
Catalog No. 11-100-8891 Supplier MilliporeSigma™ Supelco™ Supplier No. 26891U
Add to Cart
Add to Cart
Designed and tested to meet critical autosampler specifications
  • Designed with a unique plunger to eliminate bending and seizing
  • Stainless steel cone tip, point style #1 is recommended for use with pre-drilled septa
  • The shape of this syringe needle has been developed for multiple and fast septum injections on the Agilent/HP 7673A Autosampler
  • One of the most trouble-free autosampler syringes available
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Color Dark Orange
Needle Gauge 23 to 26 G
Product Type Gastight™ Syringe
Needle Point Number 1
Quantity 6 Pk.
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
Connection Type Fixed, Tapered Needle