Learn More
SGE Autosampler Syringe, SuperfleX™ Syringe, Sold by MilliporeSigma™ Supelco™
Catalog No. 111008885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
Catalog No. Volume (Metric)
11-100-8885 10 μL
1 options
Catalog No. 11-100-8885 Supplier MilliporeSigma Supelco Supplier No. 23972
Add to Cart
Add to Cart
For use with Agilent/HP 7673 and 7683 Autosamplers
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 23 G
Product Type SuperfleX™ Syringe
Needle Point Number 1
Quantity 6 Pk.
Volume (Metric) 10 μL
Product Line Superflex
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent/HP 7673 and 7683 Autosamplers
Connection Type Cemented
Show More Show Less
SuperfleX is a trademark of SGE Analytical Science Pty Ltd.