Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SGE Autosampler Syringes, Cemented Needle, 10μL

Catalog No. 111011160 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Gauge:
23 G
26 G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
11-101-1160 23 G
11-100-9315 26 G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1160 Supplier MilliporeSigma™ Supelco™ Supplier No. 29627U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with CTC/LEAP Autosamplers

TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel (Tip)
Color Dark Orange
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type Gastight™ Syringe
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) CTC, LEAP
Connection Type Cemented
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.