Description
- Maximum inertness for liquid or gas samples
- Suitable for manual disperser/diluter applications
- High accuracy of dispensed volume
- Wide variety of terminations
- Less time consuming
- Improved work flow analysis
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass (Barrel), PTFE (Plunger) |
| Product Type | SGE Gas Tight Syringe |
| Model | 25 MDR-LL-GT |
| Quantity | 1 Ea. |
| Volume (Metric) | 25 mL |
| Connection Type | Luer Lock, Kel-F Luer Cones |