Learn More
MilliporeSigma™ Supelco™ SGE Gas Tight Syringes, Luer Lock
Catalog No. 111016042 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-6042 5 mL
11-101-1184 50 μL
11-101-1194 100 μL
11-100-8872 100 mL
11-101-1140 250 μL
11-101-1127 500 μL
11-101-1002 1 mL
11-101-1041 2.5 mL
11-101-0853 10 mL
11-100-9658 25 mL
11-100-9425 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 11-101-6042 Supplier MilliporeSigma™ Supelco™ Supplier No. 21965U
Add to Cart
Add to Cart
Can be effectively used for the introduction of samples to the gas sampling and injection system (GASIS) inlet
  • Maximum inertness for liquid or gas samples
  • Suitable for manual disperser/diluter applications
  • High accuracy of dispensed volume
  • Wide variety of terminations
  • Less time consuming
  • Improved work flow analysis
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Product Type SGE Gas Tight Syringe
Model 5 MDR-LL-GT
Quantity 1 Ea.
Volume (Metric) 5 mL
Connection Type Luer Lock, Kel-F Luer Cones