Learn More
SGE Gastight™ Syringe, Removable Needle (RN), Sold by MilliporeSigma™ Supelco™
Catalog No. 111011126 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
2.5 mL
5 mL
10 mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-1126 50 μL
11-101-1115 10 μL
11-101-1117 25 μL
11-101-1012 100 μL
11-101-0993 250 μL
11-101-0994 500 μL
11-101-0805 1 mL
11-101-0830 2.5 mL
11-101-0662 5 mL
11-101-0487 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 11-101-1126 Supplier MilliporeSigma Supelco Supplier No. 26263
Add to Cart
Add to Cart
Suitable for manual dispenser/diluter applications. Maximum inertness for liquid or gas samples.
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip)
Needle Gauge 25 G
Diameter (Metric) Outer Needle 0.50 mm
Product Type SGE Gas Tight Syringe
Model 50 R-GT
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 50 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Kel-F Luer Cones, Removable
Show More Show Less
GASTIGHT is a registered trademark of Hamilton Co.