Description
- Suitable for Agilent GC
- Temperature stable up to 300°C
Specifications
Specifications
| For Use With (Application) | Viton O-Ring |
| For Use With (Equipment) | Agilent, Thermo trace 1300 Bruker (Varian) Except SG092235 |
| Material | Viton |
| Quantity | 10/Pk. |
| Type | Viton O-ring for Agilent GC's |