Learn More
MilliporeSigma™ Supelco™ SGE Guided Plunger Syringes
Catalog No. 111010837 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
Connection Type:
Fixed
Removable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
11-101-0837 5 μL Fixed
11-101-0644 5 μL Removable
11-101-0558 10 μL Removable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-0837 Supplier MilliporeSigma™ Supelco™ Supplier No. 21521
Add to Cart
Add to Cart
Extended barrel guides plunger during injection
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 26 G
Diameter (Metric) Outer Needle 0.47 mm
Product Type SGE Guided Plunger Syringe
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 5 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Fixed