Learn More
Trajan™ PTFE Seals for MOVP Valves
SDP
Catalog No. SG123687 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
MOVP Pneumatic Valves
Catalog No. For Use With (Equipment)
SG-123687 MOVP Pneumatic Valves
1 options
Catalog No. SG-123687 Supplier Trajan™ Supplier No. 123687
MOVP and MOVPT valves have pneumatically actuated on/off function.
  • Ideal for chromatography applications
  • May be used at elevated temperatures from high vacuum to medium pressure
  • Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
Type Seal
Material PTFE
For Use With (Equipment) MOVP Pneumatic Valves