Learn More
MilliporeSigma™ Supelco™ SGE Needles (25-500 μL)
Catalog No. 111011528 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
25 to 500 μL
Catalog No. Volume (Metric)
11-101-1528 25 to 500 μL
1 options
Catalog No. 11-101-1528 Supplier MilliporeSigma™ Supelco™ Supplier No. 24447
Add to Cart
Add to Cart
For use with RN Syringes (SGE 25R)
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description N25/500-5
Needle Gauge 25 G
Diameter (Metric) Outer Needle 0.50 mm
Product Type SGE Needles
Needle Point Number 2
Quantity 5 Pk.
Volume (Metric) 25 to 500 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
For Use With (Equipment) Removable Needle Syringe (SGE 25 R)