Learn More
MilliporeSigma™ Supelco™ SGE Needles for Luer Lock Syringes
Catalog No. 111011671 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Gauge:
22 G
Catalog No. Needle Gauge
11-101-1671 22 G
1 options
Catalog No. 11-101-1671 Supplier MilliporeSigma™ Supelco™ Supplier No. 26274
Add to Cart
Add to Cart
A 90° square tip with rounded and polished edges. Used for LC and HPLC valve injection and general liquid dispensing.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description NLL-LC
Needle Gauge 22 G
Diameter (Metric) Outer Needle 0.71 mm
Product Type SGE Needles
Needle Point Number 3
Quantity 2 Pk.
Length (Metric) Needle 51 mm
Needle Point Style Blunt
Connection Type Luer Lock