Learn More
MilliporeSigma™ Supelco™ SGE Needles for RN Syringes (Bevel Tip)
Catalog No. 111011527 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
Needle Gauge:
26 G
Catalog No. Volume (Metric) Needle Gauge
11-101-1527 10 μL 26 G
1 options
Catalog No. 11-101-1527 Supplier MilliporeSigma™ Supelco™ Supplier No. 24437
Add to Cart
Add to Cart
Allow for easy replacement of needles that get damaged in the field. Needle is seated at zero graduation mark.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 26 G
Product Type SGE Needles
Needle Point Number 2
Quantity 5 Pk.
Volume (Metric) 10 μL
Needle Point Style Bevel
For Use With (Equipment) Removable Needle Syringe