Learn More
MilliporeSigma™ Supelco™ SGE Needles for RN Syringes (Cone Tip)
Catalog No. 111011299 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1 μL
For Use With (Equipment):
Removable Needle Syringe (1 B-7 C)
Catalog No. Volume (Metric) For Use With (Equipment)
11-101-1299 1 μL Removable Needle Syringe (1 B-7 C)
1 options
Catalog No. 11-101-1299 Supplier MilliporeSigma™ Supelco™ Supplier No. 24432
Add to Cart
Add to Cart
For easy replacement of needles that get damaged in the field. Needle is seated at zero graduation mark.
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type SGE Needles
Model 1 B-7 C
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 1 μL
Length (Metric) Needle 70 mm
Needle Point Style Cone
For Use With (Equipment) Removable Needle Syringe (1 B-7 C)