Learn More
MilliporeSigma™ Supelco™ SGE Plunger-Protected Syringe, Fixed Needle
Catalog No. 111009611 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 Pk.
Catalog No. Quantity
11-100-9611 10 Pk.
1 options
Catalog No. 11-100-9611 Supplier MilliporeSigma™ Supelco™ Supplier No. 26239
Add to Cart
Add to Cart
Extended protection tube from top of the flange to help prevent plunger bending during injection
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Description 10 F
Needle Gauge 26 G
Product Type Plunger-Protection Syringe
Needle Point Number 2
Quantity 10 Pk.
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Fixed