Learn More
MilliporeSigma™ Supelco™ SGE Plunger-Protected Syringe, Removable Needle (RN)
Catalog No. 111011125 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
5 μL
Catalog No. Volume (Metric)
11-101-1125 5 μL
1 options
Catalog No. 11-101-1125 Supplier MilliporeSigma™ Supelco™ Supplier No. 26236
Add to Cart
Add to Cart
With a Bevel Tip, Stainless Steel Needle
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 26 G
Product Type SGE Plunger-Protected Syringe
Needle Point Number 2
Quantity 1 Ea.
Volume (Metric) 5 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel
Connection Type Removable