Learn More
Description
- Replacement Luer Lock fitting
- Contains 1 Kel-F cone and 1 threaded metal adaptor
Specifications
Specifications
| Includes | 1 Kel-F luer cone and 1 threaded metal adaptor |
| Product Type | Luer Lock Fitting |
| For Use With (Equipment) | Luer Lock Syringe |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
