Description
- Replacement plungers for gas tight syringes
- Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
| Material | PTFE (Plunger) |
| Product Type | Replacement Plunger |
| For Use With (Equipment) | Diamond Manual Syringe (250μL) |