Learn More
MilliporeSigma™ Supelco™ SGE Syringe Valves
Catalog No. 111011119 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
25 to 2500 μL
Catalog No. Volume (Metric)
11-101-1119 25 to 2500 μL
1 options
Catalog No. 11-101-1119 Supplier MilliporeSigma™ Supelco™ Supplier No. 509337
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Push-pull with needle. For use with SGE 25-2500 μL Luer Tip Syringes
  • Valve allows fluid to be stored in the syringe without contamination
  • Ideal for sample storage and transportation or to pressurize samples prior to injection
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Diameter (Metric) Outer Needle 0.63 mm
Product Type Syringe Valves
Quantity 1 Ea.
Volume (Metric) 25 to 2500 μL
Length (Metric) Needle 50 mm
Needle Point Style Cone
For Use With (Equipment) SGE Luer Tip Syringes