Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SGE Syringes, 10 μL

Catalog No. 111008920 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-100-8920 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8920 Supplier MilliporeSigma™ Supelco™ Supplier No. 24421
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Varian 8035, 8100, and 8200 GC Autosamplers

TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel (Tip)
Diameter (Metric) Outer Needle 0.5 mm
Product Type SGE Syringe
Model 10 R-VA8X
Needle Point Number 5
Quantity 1 Ea.
Volume (Metric) 10 μL
Length (Metric) Needle 53 mm
Needle Point Style Cone with Side Port
For Use With (Equipment) Varian 8035, 8100 and 8200 GC Autosamplers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.