Learn More
SGE Valco ™ Microvolume Syringe, Fixed Needle, Sold by MilliporeSigma™ Supelco™
Catalog No. 111011489 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
5 μL
100 μL
250 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
11-101-1489 100 μL
11-101-1147 250 μL
11-101-1362 5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-1489 Supplier MilliporeSigma Supelco Supplier No. 509817
Add to Cart
Add to Cart
For use with Rheodyne and Valco Valves
  • Sharp and clear fused graduation
  • Extended graduation lines and wide white backing for reduced errors
  • Angled needle tip for optimum septum penetration
  • Standard, stainless steel plunger is individually fitted to its barrel
  • Extended barrel guides the plunger during injection
  • Suitable for training and student use
  • Robust
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel (Tip)
Needle Gauge 22 G
Diameter (English) Outer Needle 0.028 in.
Product Type SGE Microvolume Syringe
Model 100 F-LC
Needle Point Number 3
Quantity 1 Ea.
Volume (Metric) 100 μL
Needle Point Style Blunt
For Use With (Equipment) Rheodyne and Valco Valves
Connection Type Fixed
Show More Show Less
Valco is a registered trademark of Valco Instruments Co. Inc.