Learn More
Reprocell Stemfactor FGF-basic, Human Recombinant (50ug)
Supplier:  Reprocell 030002
Encompass
Greater than 95% by SDS-PAGE analysis. Stemfactor bFGF is shipped at room temperature. Lyophilized bFGF is stable for up to 6 months from date of receipt when stored at −20 °C to −80 °C. Reconstituted bFGF, at concentrations greater than or equal to 0.1 mg/mL, is stable for up to 3 months when stored at −20 °C and up to 6 months when stored at −80 °C.
Catalog No. NC9790074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.