Learn More
Medchemexpress LLC N-((1-methylpiperidin-4-yl)methyl)-3-(3-(trifluoromethoxy)phenyl)imidazo[1,2-b]pyridazin-6-amine | 1025065-69-3 | MFCD16659064 | 100.0% | 405.42 g·mol⁻¹ | C20H22F3N5O | 5 MG
Supplier: Medchemexpress LLC HY132875MG
SGI-1776 (free base) is a small-molecule inhibitor of the Pim family of serine/threonine kinases intended for laboratory research. It inhibits Pim-1, Pim-2, and Pim-3 with reported IC50 values of 7 nM, 363 nM, and 69 nM, respectively, and is supplied as a high-purity solid and solution for biochemical and cell-based studies. For research use only.
- Potent Pim kinase inhibition with reported IC50 values for Pim-1, Pim-2, and Pim-3.
- High purity suitable for biochemical and cell-based assays.
- Available in solid and solution formats for flexible experimental use.
- Applicable to kinase signaling, apoptosis, and cancer biology research.
- Delivered with supporting documentation such as COA and SDS for traceability.