Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGLT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584085
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584085 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584085 Supplier Invitrogen™ Supplier No. PA584085
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLDAEEENIQ EGPKETIEIE TQVPEKKKGI FRRAYDLFCG LEQHGAPKMT EEEEKAMKMK MTDTSEKPLW R Highest antigen sequence indentity to the following orthologs: Rat - 71%, Mouse - 76%.

Glucose transporters are integral membrane proteins that mediate the transport of glucose and structurally-related substances across cellular membranes. Two families of glucose transporter have been identified: the facilitated-diffusion glucose transporter family. The SLC5A1 gene encodes a protein that is involved in the active transport of glucose and galactose into eukaryotic and some prokaryotic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGLT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc5a1
Gene Accession No. P13866
Gene Alias D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter)
Gene Symbols Slc5a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC5A1. Recombinant protein control fragment (Product #RP-100900).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6523
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.