Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGMS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583764 Supplier Invitrogen™ Supplier No. PA583764
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASTMKEVVYW SPKKVADWLL ENAMPEYCEP LEHFTGQDLI NLTQEDFKKP PLCRVSSDNG QRLLDMIETL KMEHHLEAHK NGHANGHLNI GVDIPTPDGS FSIKIKPNGM PNGYRKEMIK IPM Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.

A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGMS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGMS1
Gene Accession No. Q86VZ5
Gene Alias 9530058O11Rik; AI841905; C80702; hmob33; Medulla oblongata-derived protein; MGC17342; MOB; MOB1; mob-1; Phosphatidylcholine:ceramide cholinephosphotransferase 1; protein Mob; SGMS1; SMS1; Sor1; sphingomyelin synthase 1; TMEM23; Transmembrane protein 23
Gene Symbols SGMS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGMS1. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259230
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.