Learn More
Invitrogen™ SGPL1 Polyclonal Antibody

Description
By Western blot, PA1-12722 detects a ~64 kDa protein representing S1P lyase 1 from MDBK cell lysate.
Specifications
Specifications
| Antigen | SGPL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | SGPL1 |
| Gene Accession No. | O95470, Q8CHN6, Q8R0X7 |
| Gene Alias | AI428538; D10Xrf456; FLJ13811; hSPL; KIAA1252; mSPL; S1PL; Sgpl1; Spgl1; sphingosine phosphate lyase 1; sphingosine-1-phosphate aldolase; Sphingosine-1-phosphate lyase 1; sphingosine-1-phosphate lyase 1-like protein; spl; SPL 1; SP-lyase 1; zgc:162153 |
| Gene Symbols | SGPL1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.