Learn More
Description
The SGPP1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 81537 |
| Species | Human |
| Product Line | Novus |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | SGPP1 |
| Label Type | Unconjugated |
| Molecular Weight (g/mol) | 20 kDa |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
