Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGTA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106280
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106280 Supplier Invitrogen™ Supplier No. PA5106280
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SGTA.

This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGTA
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SGTA
Gene Accession No. O43765, O70593, Q8BJU0
Gene Alias 5330427H01Rik; AI194281; alpha h UBP; alphaSGT; Alpha-SGT; D10Ertd190e; hSGT; protein containing three tetratricopeptide repeats; Sgt; Sgt1; SGTA; small glutamine rich tetratricopeptide repeat containing alpha; small glutamine-rich protein with tetratricopeptide repeats 1; small glutamine-rich tetratricopeptide repeat (TPR) containing protein; small glutamine-rich tetratricopeptide repeat (TPR) containing protein (SGT); small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha; small glutamine-rich tetratricopeptide repeat-containing protein alpha; Stg; UBP; vpu-binding protein
Gene Symbols SGTA
Host Species Rabbit
Immunogen A synthesized peptide derived from human SGTA(Accession O43765), corresponding to amino acid residues N255-S305.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52551, 6449, 64667
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.