Learn More
Apexbio Technology LLC SH-4-54,5mg CAS# 1456632-40-8
SDP
Supplier:  Apexbio Technology LLC B47895
Encompass_Preferred
SH-4-54 is a potent inhibitor of STAT3 with KD values of 300 and 464 nM for STAT3 and STAT5, respectively [1].STAT3 is a transcription factor that belongs to the STAT transcription factor family. STAT3 signaling plays an important role in the survival and proliferation of brain tumor stem cells (BTSCs) and glioblastoma (GBM) [1].SH-4-54 is a potent STAT inhibitor. SH-4-54 inhibited interactions between STAT3 and phosphopeptide with Ki value of 10-30 μM. SH-4-54 exhibited selectivity for STAT3 against STAT1 by >10-fold. SH-4-54 inhibited pSTAT3 (Y705) in a concentration-dependent way with no effect on pSTAT3 (S727), suggesting that SH-4-54 bound to the STAT3 SH2 domain. Also, SH-4-54 reduced pSTAT3 levels and inhibited the downstream targets Cyclin D1 and Bcl-xL, which were involved in cell growth and survival [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0024
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.