Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SH2D1A Antibody Pair [HRP]
SDP

Catalog No. NB451140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB451140 15 Plates
NB451139 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB451140 Supplier Novus Biologicals™ Supplier No. NBP32691615PLATES
Add to Cart
Add to Cart

Antibody Pair

specifically detects in Human samples. It is validated for ELISA∣Sandwich ELISA.

Specifications

Assay Range 34.38-2200 pg/mL (example only; lot dependent)
Assay Sensitivity 34.38 pg/mL (example only; lot dependent)
Conjugate HRP
Product Type Sandwich ELISA
Target Species Human
For Use With (Application) Sandwich ELISA
Gene ID (Entrez) 4068
Gene Symbol SH2D1A
Quantity 15 Plates
Research Discipline Immune System Diseases, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.