Learn More
Invitrogen™ SH2D4A Polyclonal Antibody

Description
Immunogen sequence: LFFKMREEQI RRWKEREAAM ERKESLPVKP RPKKENGKSV HWKLGADKEV WVWVMGEHHL DKPYDVLCNE IIAERARLKA EQEAEEPRKT HSEEFTNSLK TKSQYHDLQA PDNQQTKDIW KKVA Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.
Specifications
Specifications
| Antigen | SH2D4A |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SH2D4A |
| Gene Accession No. | Q9H788 |
| Gene Alias | 2210402M20Rik; FLJ20967; PPP1R38; protein phosphatase 1 regulatory subunit 38; protein phosphatase 1, regulatory subunit 38; protein SH(2)A; SH2 domain containing 4A; SH2 domain-containing 4A; SH2 domain-containing protein 4A; SH2A; SH2D4A |
| Gene Symbols | SH2D4A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.