Learn More
Specifications
Specifications
| Gene ID (Entrez) | 646892 |
| Species | Human |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-17142It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.For current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here . |
| Gene Alias | SH2D7 SH2 domain containing 7 |
| Gene Symbol | SH2D7 |
| Molecular Weight (g/mol) | 27 kDa |
| Quantity | 0.1 mL |
| Storage Buffer | PBS and 1M Urea, pH 7.4. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
