Learn More
Description
A suggested positive control for this product is recombinant 3BP2 protein.
Specifications
Specifications
| Antigen | SH3BP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Sh3bp2 |
| Gene Accession No. | Q06649 |
| Gene Alias | 3BP2; 3BP-2; Abl-SH3 binding protein 2; CRBM; CRPM; FLJ42079; LOW QUALITY PROTEIN: SH3 domain-binding protein 2; RES4-23; SH3 domain binding protein 2; SH3 domain-binding protein 2; SH3BP2; SH3-domain binding protein 2; TNFAIP3 interacting protein 2 |
| Gene Symbols | Sh3bp2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
