Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501735533 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5533 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5533 Supplier Proteintech Group Inc Supplier No. 270141AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Implicated in endocytosis. May recruit other proteins to membranes with high curvature.

Specifications

Antigen SH3GL1
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SH3GL1
Gene Accession No. O35964, Q62419, Q99961
Gene Alias CNSA1, EEN, EEN fusion partner of MLL, Endophilin 2, Endophilin A2, SH3 domain GRB2 like 1, SH3 domain protein 2B, SH3D2B, SH3GL1, SH3P8
Gene Symbols SH3GL1
Host Species Rabbit
Immunogen SH3GL1 Fusion Protein Ag25550
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20405, 6455, 81922
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.